Assay Details
| Gene Symbol: hamp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097260.1 | NP_001090729.1 | 1 - 2 | 109 | 94 | |
| GenBank mRNA | BC156006.1 | - | 1 - 2 | 106 | 94 | |
| EF140888.1 | - | 1 - 2 | 109 | 94 | ||
More Information
Set Membership: |
|

hamp
Genomic Map