Assay Details
| Gene Symbol: hsd11b1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097315.1 | NP_001090784.1 | 6 - 7 | 757 | 69 | |
| GenBank mRNA | BC135167.1 | - | 6 - 7 | 757 | 69 | |
More Information
Set Membership: |
|

hsd11b1l
Genomic Map