Assay Details
| Gene Symbol: caps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097320.1 | NP_001090789.1 | 2 - 3 | 210 | 97 | |
| XM_012957609.2 | XP_012813063.1 | 3 - 4 | 269 | 97 | ||
| XM_018091169.1 | XP_017946658.1 | 3 - 4 | 269 | 97 | ||
| XM_018091179.1 | XP_017946668.1 | 2 - 3 | 747 | 97 | ||
| XM_018091197.1 | XP_017946686.1 | 3 - 4 | 509 | 97 | ||
| XM_018091214.1 | XP_017946703.1 | 2 - 3 | 412 | 97 | ||
| XM_018091227.1 | XP_017946716.1 | 2 - 3 | 434 | 97 | ||
| XM_018091240.1 | XP_017946729.1 | 2 - 3 | 343 | 97 | ||
| GenBank mRNA | BC135179.1 | - | 2 - 3 | 210 | 97 | |
More Information
Set Membership: |
|

caps
Genomic Map