Assay Details
Gene Symbol: rnls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097322.1 | NP_001090791.1 | 1 - 2 | 130 | 85 | |
XM_012965827.2 | XP_012821281.1 | 1 - 2 | 241 | 85 | ||
GenBank mRNA | BC135183.1 | - | 1 - 2 | 130 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
