Assay Details
Gene Symbol: tuba8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097330.1 | NP_001090799.1 | 465 | 55 | ||
GenBank mRNA | BC135216.1 | - | 465 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
