Assay Details
Gene Symbol: epha10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097348.1 | NP_001090817.1 | 1 - 2 | 121 | 76 | |
GenBank mRNA | BC135268.1 | - | 1 - 2 | 121 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
