Assay Details
| Gene Symbol: heca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097357.1 | NP_001090826.1 | 2 - 3 | 1472 | 89 | |
| XM_012962552.2 | XP_012818006.1 | 2 - 3 | 2181 | 89 | ||
| XM_018093737.1 | XP_017949226.1 | 2 - 3 | 2181 | 89 | ||
| GenBank mRNA | BC089663.1 | - | 3 - 4 | 1600 | 89 | |
| BC135464.1 | - | 2 - 3 | 1472 | 89 | ||
More Information
Set Membership: |
|

heca
Genomic Map