Assay Details
| Gene Symbol: btg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097371.1 | NP_001090840.1 | 4 - 5 | 623 | 59 | |
| GenBank mRNA | BC118746.1 | - | 4 - 5 | 623 | 59 | |
| CT025239.2 | - | 4 - 5 | 576 | 59 | ||
More Information
Set Membership: |
|

btg3
Genomic Map