Assay Details
| Gene Symbol: rfx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097380.1 | NP_001090849.1 | 3 - 4 | 707 | 64 | |
| XM_012958367.2 | XP_012813821.1 | 2 - 3 | 985 | 64 | ||
| XM_012958368.2 | XP_012813822.1 | 3 - 4 | 812 | 64 | ||
| GenBank mRNA | BC133720.1 | - | 3 - 4 | 707 | 64 | |
| CR761154.2 | - | 3 - 4 | 472 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_012958368.2 transcripts.
Set Membership: |
|

rfx7
Genomic Map