Assay Details
| Gene Symbol: slc25a15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097397.1 | NP_001090866.1 | 3 - 4 | 549 | 66 | |
| XM_012956239.2 | XP_012811693.1 | 4 - 5 | 664 | 66 | ||
| GenBank mRNA | BC135300.1 | - | 3 - 4 | 549 | 66 | |
More Information
Set Membership: |
|

slc25a15
Genomic Map