Assay Details
Gene Symbol: gk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097398.1 | NP_001090867.1 | 12 - 13 | 1013 | 61 | |
XM_012956242.2 | XP_012811696.1 | 13 - 14 | 1167 | 61 | ||
XM_012956243.2 | XP_012811697.1 | 13 - 14 | 1168 | 61 | ||
GenBank mRNA | BC135303.1 | - | 12 - 13 | 1013 | 61 | |
CT025212.2 | - | 13 - 14 | 1107 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
