Assay Details
| Gene Symbol: snca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097407.1 | NP_001090876.1 | 5 - 6 | 702 | 82 | |
| XM_012959451.2 | XP_012814905.1 | 5 - 6 | 672 | 82 | ||
| XM_018091927.1 | XP_017947416.1 | 5 - 6 | 803 | 82 | ||
| GenBank mRNA | BC135333.1 | - | 5 - 6 | 702 | 82 | |
More Information
Set Membership: |
|

snca
Genomic Map