Assay Details
| Gene Symbol: snx15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100197.1 | NP_001093667.1 | 3 - 4 | 286 | 72 | |
| GenBank mRNA | BC135952.1 | - | 3 - 4 | 286 | 72 | |
| CR942341.2 | - | 3 - 4 | 249 | 72 | ||
More Information
Set Membership: |
|

snx15
Genomic Map