Assay Details
| Gene Symbol: nr1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100205.1 | NP_001093675.1 | 7 - 8 | 1724 | 75 | |
| GenBank mRNA | BC135990.1 | - | 7 - 8 | 1724 | 75 | |
More Information
Set Membership: |
|

nr1d1
Genomic Map