Assay Details
| Gene Symbol: edil3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100213.1 | NP_001093683.1 | 6 - 7 | 938 | 73 | |
| GenBank mRNA | BC136021.1 | - | 6 - 7 | 938 | 73 | |
More Information
Set Membership: |
|

edil3
Genomic Map