Assay Details
Gene Symbol: dynlrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100217.1 | NP_001093687.1 | 3 - 4 | 266 | 83 | |
XM_012971496.2 | XP_012826950.1 | 3 - 4 | 327 | 83 | ||
GenBank mRNA | BC136095.1 | - | 3 - 4 | 266 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
