Assay Details
Gene Symbol: shox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100224.1 | NP_001093694.1 | 2 - 3 | 653 | 62 | |
XM_012962582.2 | XP_012818036.1 | 2 - 3 | 893 | 62 | ||
GenBank mRNA | DQ675518.1 | - | 2 - 3 | 653 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
