Assay Details
| Gene Symbol: rac3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100242.1 | NP_001093712.1 | 5 - 6 | 529 | 68 | |
| GenBank mRNA | BC135751.1 | - | 5 - 6 | 529 | 68 | |
More Information
Set Membership: |
|

rac3
Genomic Map