Assay Details
| Gene Symbol: poll | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100246.1 | NP_001093716.1 | 6 - 7 | 1053 | 63 | |
| XM_012965851.2 | XP_012821305.1 | 6 - 7 | 1105 | 63 | ||
| GenBank mRNA | BC135821.1 | - | 6 - 7 | 1053 | 63 | |
More Information
Set Membership: |
|

poll
Genomic Map