Assay Details
Gene Symbol: unc93b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100253.1 | NP_001093723.1 | 5 - 6 | 673 | 85 | |
XM_012960408.2 | XP_012815862.1 | 5 - 6 | 719 | 85 | ||
GenBank mRNA | BC135524.1 | - | 5 - 6 | 673 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
