Assay Details
Gene Symbol: tmeff1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100268.3 | NP_001093738.2 | 1 - 2 | 682 | 65 | |
XM_012964170.2 | XP_012819624.1 | 1 - 2 | 732 | 65 | ||
GenBank mRNA | BC135422.2 | - | 1 - 2 | 516 | 65 | |
CX391920.2 | - | 1 - 2 | 680 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
