Assay Details
Gene Symbol: med19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101806.1 | NP_001095276.1 | 3 - 4 | 497 | 68 | |
GenBank mRNA | BC088543.1 | - | 3 - 4 | 488 | 68 | |
BC160502.1 | - | 3 - 4 | 549 | 68 | ||
CR942536.2 | - | 3 - 4 | 497 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
