Assay Details
| Gene Symbol: lats2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102704.1 | NP_001096174.1 | 3 - 4 | 824 | 63 | |
| XM_012956265.2 | XP_012811719.1 | 3 - 4 | 1162 | 63 | ||
| XM_012956266.2 | XP_012811720.1 | 3 - 4 | 814 | 63 | ||
| XM_012956267.2 | XP_012811721.1 | 3 - 4 | 1160 | 63 | ||
| GenBank mRNA | BC136036.1 | - | 3 - 4 | 824 | 63 | |
More Information
Set Membership: |
|

lats2
Genomic Map