Assay Details
| Gene Symbol: pcdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102705.1 | NP_001096175.1 | 2 - 3 | 1052 | 89 | |
| XM_012958411.2 | XP_012813865.1 | 2 - 3 | 1834 | 89 | ||
| XM_012958412.1 | XP_012813866.1 | 2 - 3 | 852 | 89 | ||
| XM_012958415.2 | XP_012813869.1 | 3 - 4 | 1335 | 89 | ||
| XM_012958416.1 | XP_012813870.1 | 2 - 3 | 932 | 89 | ||
| XM_012958417.2 | XP_012813871.1 | 2 - 3 | 1215 | 89 | ||
| XM_012958418.1 | XP_012813872.1 | 2 - 3 | 859 | 89 | ||
| XM_018091905.1 | XP_017947394.1 | 3 - 4 | 1011 | 89 | ||
| XM_018091906.1 | XP_017947395.1 | 2 - 3 | 1836 | 89 | ||
| XM_018091907.1 | XP_017947396.1 | 2 - 3 | 1835 | 89 | ||
| GenBank mRNA | BC135899.1 | - | 2 - 3 | 1052 | 89 | |
| CT027868.1 | - | 2 - 3 | 864 | 89 | ||
More Information
Set Membership: |
|

pcdh1
Genomic Map