Assay Details
| Gene Symbol: gpr78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102723.1 | NP_001096193.1 | 2 - 3 | 1186 | 74 | |
| GenBank mRNA | BC136127.1 | - | 2 - 3 | 1186 | 74 | |
More Information
Set Membership: |
|

gpr78
Genomic Map