Assay Details
| Gene Symbol: dlc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102772.1 | NP_001096242.1 | 2 - 3 | 731 | 76 | |
| GenBank mRNA | BC135385.1 | - | 2 - 3 | 731 | 76 | |
More Information
Set Membership: |
|

dlc
Genomic Map