Assay Details
| Gene Symbol: kyat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102788.1 | NP_001096258.1 | 8 - 9 | 1725 | 60 | |
| GenBank mRNA | BC084158.1 | - | 8 - 9 | 804 | 60 | |
| BC135439.1 | - | 8 - 9 | 1725 | 60 | ||
More Information
Set Membership: |
|

kyat3
Genomic Map