Assay Details
Gene Symbol: hm13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102789.1 | NP_001096259.1 | 326 | 77 | ||
GenBank mRNA | BC135444.1 | - | 326 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
