Assay Details
| Gene Symbol: tyw1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102797.2 | NP_001096267.1 | 2083 | 63 | ||
| GenBank mRNA | BC135483.1 | - | 2085 | 63 | ||
| CT030473.1 | - | 14 - 15 | 1939 | 63 | ||
More Information
Set Membership: |
|

tyw1
Genomic Map