Assay Details
Gene Symbol: blvrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102799.1 | NP_001096269.1 | 4 - 5 | 462 | 66 | |
XM_012967878.1 | XP_012823332.1 | 4 - 5 | 550 | 66 | ||
GenBank mRNA | BC135495.1 | - | 4 - 5 | 462 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
