Assay Details
Gene Symbol: c3orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102804.1 | NP_001096274.1 | 3 - 4 | 709 | 78 | |
GenBank mRNA | BC135506.1 | - | 3 - 4 | 709 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
