Assay Details
Gene Symbol: psma5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102812.1 | NP_001096282.1 | 2 - 3 | 207 | 86 | |
GenBank mRNA | BC135540.1 | - | 2 - 3 | 207 | 86 | |
CT485761.1 | - | 2 - 3 | 197 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
