Assay Details
Gene Symbol: rnaseh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102829.1 | NP_001096299.1 | 4 - 5 | 603 | 72 | |
XM_012962617.2 | XP_012818071.1 | 4 - 5 | 832 | 72 | ||
XM_018093760.1 | XP_017949249.1 | 4 - 5 | 699 | 72 | ||
GenBank mRNA | BC135622.1 | - | 4 - 5 | 603 | 72 | |
CT025221.2 | - | 4 - 5 | 518 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
