Assay Details
| Gene Symbol: lgals9c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102848.1 | NP_001096318.1 | 2 - 3 | 119 | 82 | |
| XM_012956288.2 | XP_012811742.1 | 2 - 3 | 202 | 82 | ||
| XM_012956289.2 | XP_012811743.1 | 2 - 3 | 203 | 82 | ||
| XM_012956290.2 | XP_012811744.1 | 2 - 3 | 203 | 82 | ||
| GenBank mRNA | BC081298.1 | - | 2 - 3 | 157 | 82 | |
| BC135696.1 | - | 2 - 3 | 119 | 82 | ||
More Information
Set Membership: |
|

lgals9c
Genomic Map