Assay Details
Gene Symbol: nat8l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102880.1 | NP_001096350.1 | 2 - 3 | 618 | 67 | |
GenBank mRNA | BC135824.1 | - | 2 - 3 | 618 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
