Assay Details
| Gene Symbol: zc2hc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102891.1 | NP_001096361.1 | 5 - 6 | 565 | 76 | |
| XM_012964190.2 | XP_012819644.1 | 5 - 6 | 1374 | 76 | ||
| GenBank mRNA | BC135865.1 | - | 5 - 6 | 565 | 76 | |
More Information
Set Membership: |
|

zc2hc1a
Genomic Map