Assay Details
| Gene Symbol: brms1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102896.2 | NP_001096366.1 | 4 - 5 | 497 | 60 | |
| GenBank mRNA | BC075612.1 | - | 4 - 5 | 500 | 60 | |
| BC135888.1 | - | 4 - 5 | 502 | 60 | ||
More Information
Set Membership: |
|

brms1l
Genomic Map