Assay Details
| Gene Symbol: cnot7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102905.1 | NP_001096375.1 | 2 - 3 | 370 | 79 | |
| XM_018093877.1 | XP_017949366.1 | 2 - 3 | 812 | 79 | ||
| GenBank mRNA | BC135920.1 | - | 2 - 3 | 370 | 79 | |
More Information
Set Membership: |
|

cnot7
Genomic Map