Assay Details
Gene Symbol: rab7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102911.1 | NP_001096381.1 | 4 - 5 | 608 | 97 | |
XM_012965077.2 | XP_012820531.1 | 4 - 5 | 807 | 97 | ||
GenBank mRNA | BC135955.1 | - | 4 - 5 | 608 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
