Assay Details
| Gene Symbol: ormdl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102917.1 | NP_001096387.1 | 3 - 4 | 399 | 82 | |
| XM_012969979.2 | XP_012825433.1 | 4 - 5 | 478 | 82 | ||
| GenBank mRNA | BC135976.1 | - | 3 - 4 | 399 | 82 | |
More Information
Set Membership: |
|

ormdl1
Genomic Map