Assay Details
| Gene Symbol: samd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102935.1 | NP_001096405.1 | 5 - 6 | 640 | 60 | |
| XM_012965901.2 | XP_012821355.1 | 10 - 11 | 2411 | 60 | ||
| XM_012965902.2 | XP_012821356.1 | 10 - 11 | 2360 | 60 | ||
| XM_012965903.2 | XP_012821357.1 | 9 - 10 | 2312 | 60 | ||
| XM_012965904.2 | XP_012821358.1 | 10 - 11 | 2424 | 60 | ||
| XM_012965906.2 | XP_012821360.1 | 10 - 11 | 1170 | 60 | ||
| XM_012965908.2 | XP_012821362.1 | 10 - 11 | 1119 | 60 | ||
| XM_012965909.2 | XP_012821363.1 | 10 - 11 | 1100 | 60 | ||
| XM_012965910.2 | XP_012821364.1 | 10 - 11 | 1107 | 60 | ||
| XM_012965911.2 | XP_012821365.1 | 10 - 11 | 1100 | 60 | ||
| XM_012965912.2 | XP_012821366.1 | 10 - 11 | 1040 | 60 | ||
| XM_012965913.2 | XP_012821367.1 | 10 - 11 | 1050 | 60 | ||
| XM_012965914.2 | XP_012821368.1 | 9 - 10 | 999 | 60 | ||
| GenBank mRNA | BC136045.1 | - | 5 - 6 | 640 | 60 | |
| CR942606.2 | - | 5 - 6 | 655 | 60 | ||
More Information
Set Membership: |
|

samd11
Genomic Map