Assay Details
Gene Symbol: adcyap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102955.1 | NP_001096425.1 | 2 - 3 | 473 | 70 | |
XM_012964194.2 | XP_012819648.1 | 2 - 3 | 936 | 70 | ||
XM_012964195.2 | XP_012819649.1 | 2 - 3 | 822 | 70 | ||
XM_012964196.2 | XP_012819650.1 | 2 - 3 | 932 | 70 | ||
GenBank mRNA | BC136094.1 | - | 2 - 3 | 473 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
