Assay Details
Gene Symbol: ubash3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102961.1 | NP_001096431.1 | 1 - 2 | 384 | 59 | |
GenBank mRNA | BC136111.1 | - | 1 - 2 | 384 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
