Assay Details
| Gene Symbol: orai1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102978.1 | NP_001096448.1 | 2 - 3 | 350 | 86 | |
| XM_018091046.1 | XP_017946535.1 | 2 - 3 | 2263 | 86 | ||
| GenBank mRNA | BC136155.1 | - | 2 - 3 | 350 | 86 | |
More Information
Set Membership: |
|

orai1
Genomic Map