Assay Details
| Gene Symbol: baat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102991.1 | NP_001096461.1 | 4 - 5 | 794 | 86 | |
| GenBank mRNA | BC136191.1 | - | 4 - 5 | 794 | 86 | |
More Information
Set Membership: |
|

baat
Genomic Map