Assay Details
| Gene Symbol: mettl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103013.1 | NP_001096483.1 | 6 - 7 | 996 | 91 | |
| XM_012964209.2 | XP_012819663.1 | 6 - 7 | 1201 | 91 | ||
| XM_012964210.2 | XP_012819664.1 | 5 - 6 | 1046 | 91 | ||
| XM_012964211.2 | XP_012819665.1 | 5 - 6 | 923 | 91 | ||
| XM_018094516.1 | XP_017950005.1 | 5 - 6 | 778 | 91 | ||
| XM_018094517.1 | XP_017950006.1 | 6 - 7 | 1193 | 91 | ||
| GenBank mRNA | BC136247.1 | - | 6 - 7 | 996 | 91 | |
| BC155866.1 | - | 5 - 6 | 937 | 91 | ||
More Information
Set Membership: |
|

mettl6
Genomic Map