Assay Details
| Gene Symbol: rps15a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103019.1 | NP_001096489.1 | 3 - 4 | 322 | 76 | |
| GenBank mRNA | BC135393.1 | - | 3 - 4 | 322 | 76 | |
More Information
Set Membership: |
|

rps15a
Genomic Map