Assay Details
Gene Symbol: ndufa8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103021.1 | NP_001096491.1 | 1 - 2 | 65 | 87 | |
GenBank mRNA | BC135449.1 | - | 1 - 2 | 65 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
