Assay Details
| Gene Symbol: fkbp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103022.1 | NP_001096492.1 | 2 - 3 | 128 | 65 | |
| GenBank mRNA | BC135491.1 | - | 2 - 3 | 128 | 65 | |
More Information
Set Membership: |
|

fkbp1b
Genomic Map