Assay Details
| Gene Symbol: wasf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103026.1 | NP_001096496.1 | 5 - 6 | 563 | 95 | |
| XM_018091050.1 | XP_017946539.1 | 5 - 6 | 567 | 95 | ||
| GenBank mRNA | BC135144.1 | - | 5 - 6 | 563 | 95 | |
More Information
Set Membership: |
|

wasf2
Genomic Map